Product Details

SNP ID
rs41264495
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:39624371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGAGCTGGATCATTCCCATCT[A/C]ATTTCAGTGGCATCACAGATTCTTT
Phenotype
MIM: 609034
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HEYL PubMed Links

Gene Details

Gene
HEYL
Gene Name
hes related family bHLH transcription factor with YRPW motif-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014571.3 3174 UTR 3 NP_055386.1
XM_005270745.2 3174 UTR 3 XP_005270802.1

View Full Product Details