Product Details

SNP ID
rs41267037
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:39760431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCGGGTGGACTCAGTGGCAGCC[A/G]CAGGCCTTGACCACCATGTTGCGGT
Phenotype
MIM: 602284 MIM: 610289 MIM: 602435
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMP8B PubMed Links

Gene Details

Gene
BMP8B
Gene Name
bone morphogenetic protein 8b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005271149.4 1366 Intron XP_005271206.1
XM_011542022.2 1366 Intron XP_011540324.1
XM_011542024.2 1366 Silent Mutation XP_011540326.2
XM_011542025.2 1366 Intron XP_011540327.1
XM_017002155.1 1366 Intron XP_016857644.1
XM_017002156.1 1366 Intron XP_016857645.1
Gene
OXCT2
Gene Name
3-oxoacid CoA-transferase 2
There are no transcripts associated with this gene.

Gene
PPIE
Gene Name
peptidylprolyl isomerase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195007.1 1366 Intron NP_001181936.1
NM_001319293.1 1366 Intron NP_001306222.1
NM_006112.3 1366 Intron NP_006103.1
NM_203456.2 1366 Intron NP_982281.1
XM_006710289.2 1366 Intron XP_006710352.1
XM_006710290.3 1366 Intron XP_006710353.1
XM_011540501.1 1366 Intron XP_011538803.1
XM_017000051.1 1366 Intron XP_016855540.1
XM_017000052.1 1366 Intron XP_016855541.1

View Full Product Details