Product Details

SNP ID
rs41270389
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:43970227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGCGGCGCTGCAGCGAGAGACC[A/G]GGGTGCCAGGACTGCTTACTCCTCT
Phenotype
MIM: 603717 MIM: 604013 MIM: 603456 MIM: 610411
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0B PubMed Links

Gene Details

Gene
ATP6V0B
Gene Name
ATPase H+ transporting V0 subunit b
There are no transcripts associated with this gene.

Gene
B4GALT2
Gene Name
beta-1,4-galactosyltransferase 2
There are no transcripts associated with this gene.

Gene
DPH2
Gene Name
DPH2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039589.1 247 Missense Mutation AGG,GGG R18G NP_001034678.1
NM_001319165.1 247 UTR 5 NP_001306094.1
NM_001319166.1 247 Missense Mutation AGG,GGG R18G NP_001306095.1
NM_001319167.1 247 UTR 5 NP_001306096.1
NM_001319168.1 247 UTR 5 NP_001306097.1
NM_001319169.1 247 UTR 5 NP_001306098.1
NM_001319170.1 247 UTR 5 NP_001306099.1
NM_001319171.1 247 UTR 5 NP_001306100.1
NM_001384.4 247 Missense Mutation AGG,GGG R18G NP_001375.2
XM_017000502.1 247 Missense Mutation AGG,GGG R18G XP_016855991.1
XM_017000503.1 247 UTR 5 XP_016855992.1
XM_017000504.1 247 UTR 5 XP_016855993.1
XM_017000505.1 247 UTR 5 XP_016855994.1
XM_017000506.1 247 UTR 5 XP_016855995.1
Gene
IPO13
Gene Name
importin 13
There are no transcripts associated with this gene.

View Full Product Details