Product Details

SNP ID
rs41287260
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:100266848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTCTCGAGTTTTCTCTGTTTG[A/G]CTAAATAGTGTCATTTGCTGAAGAG
Phenotype
MIM: 611286
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTCA PubMed Links

Gene Details

Gene
RTCA
Gene Name
RNA 3'-terminal phosphate cyclase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130841.1 Intron NP_001124313.1
NM_003729.3 Intron NP_003720.1
XM_005271298.4 Intron XP_005271355.1
XM_005271299.4 Intron XP_005271356.1
XM_017002669.1 Intron XP_016858158.1
Gene
RTCA-AS1
Gene Name
RTCA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details