Product Details
- SNP ID
-
rs45521433
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118978627 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAGAGGGGAATGAGAAGGTCAGA[A/G]AGGTAAAGGGAAAACCCGTAGAAGT
- Phenotype
-
MIM: 615755
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXR1
PubMed Links
Gene Details
- Gene
- FOXR1
- Gene Name
- forkhead box R1
View Full Product Details