Product Details

SNP ID
rs41295125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:6091802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCGCTGTGTGAAACAATAGGCAG[C/T]ATAACTTGCCAAGGTTGCTGTGATC
Phenotype
MIM: 606935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RBM17 PubMed Links

Gene Details

Gene
RBM17
Gene Name
RNA binding motif protein 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145547.1 Intron NP_001139019.1
NM_032905.4 Intron NP_116294.1

View Full Product Details