Product Details

SNP ID
rs41312118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48898207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGCTGGATGAGCCCATTTCTGAA[G/T]TGCGGAGGGGCCGGGCTTCTTAGGC
Phenotype
MIM: 300463 MIM: 314375 MIM: 300249
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PQBP1 PubMed Links

Gene Details

Gene
PQBP1
Gene Name
polyglutamine binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032381.1 98 Intron NP_001027553.1
NM_001032382.1 98 Intron NP_001027554.1
NM_001032383.1 98 Intron NP_001027555.1
NM_001032384.1 98 Intron NP_001027556.1
NM_001167989.1 98 Intron NP_001161461.1
NM_001167990.1 98 Intron NP_001161462.1
NM_001167992.1 98 Intron NP_001161464.1
NM_005710.2 98 Intron NP_005701.1
NM_144495.2 98 Intron NP_652766.1
XM_005272571.3 98 Intron XP_005272628.1
XM_005272572.4 98 Intron XP_005272629.1
XM_011543884.2 98 UTR 5 XP_011542186.1
XM_017029207.1 98 Intron XP_016884696.1
Gene
SLC35A2
Gene Name
solute carrier family 35 member A2
There are no transcripts associated with this gene.

Gene
TIMM17B
Gene Name
translocase of inner mitochondrial membrane 17 homolog B (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167947.2 98 Intron NP_001161419.1
NM_005834.4 98 Intron NP_005825.1

View Full Product Details