Product Details

SNP ID
hCV88322604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:1435814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCGCTGGCGCGGAGCGGCGC[A/G]GAGCGCGGTGAGTGCGGCGGGCGGC
Phenotype
MIM: 611901
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724312 PubMed Links

Gene Details

Gene
LOC102724312
Gene Name
uncharacterized LOC102724312
There are no transcripts associated with this gene.

Gene
LOC107985729
Gene Name
uncharacterized LOC107985729
There are no transcripts associated with this gene.

Gene
TMEM88B
Gene Name
transmembrane protein 88B
There are no transcripts associated with this gene.

Gene
VWA1
Gene Name
von Willebrand factor A domain containing 1
There are no transcripts associated with this gene.

View Full Product Details