Product Details

SNP ID
rs61857829
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247712045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGATGCGAGGAGCACGTGGAGAA[G/T]GCTTTGCTCCGGCCACTGGCAGAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 711 Silent Mutation GCA,GCC A237A NP_001005286.1

View Full Product Details