Product Details
- SNP ID
-
rs62182739
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:177624255 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTTAATGAATCCTTTTTGTGTTT[G/T]GGTGGTTTTTTTTTTTTTCAGCCAA
- Phenotype
-
MIM: 604961
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PDE11A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4020005] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDE11A
- Gene Name
- phosphodiesterase 11A
- Gene
- TTC30A
- Gene Name
- tetratricopeptide repeat domain 30A
There are no transcripts associated with this gene.
View Full Product Details