Product Details

SNP ID
rs61746229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178441670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGGCAAGAAATTTCTCAGCAGC[A/G]TTCCTTTTGGCTTGCTTTTTTGATG
Phenotype
MIM: 610219 MIM: 603424
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DFNB59 PubMed Links

Gene Details

Gene
DFNB59
Gene Name
deafness, autosomal recessive 59
There are no transcripts associated with this gene.

Gene
LOC101927027
Gene Name
uncharacterized LOC101927027
There are no transcripts associated with this gene.

Gene
PRKRA
Gene Name
protein activator of interferon induced protein kinase EIF2AK2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139517.1 791 Silent Mutation AAC,AAT N172N NP_001132989.1
NM_001139518.1 791 Silent Mutation AAC,AAT N158N NP_001132990.1
NM_001316362.1 791 Silent Mutation AAC,AAT N70N NP_001303291.1
NM_003690.4 791 Silent Mutation AAC,AAT N183N NP_003681.1
XM_011512063.1 791 Silent Mutation AAC,AAT N98N XP_011510365.1
XM_011512066.1 791 Silent Mutation AAC,AAT N70N XP_011510368.1
XM_017005159.1 791 Silent Mutation AAC,AAT N70N XP_016860648.1

View Full Product Details