Product Details

SNP ID
rs55737555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201077196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATATTTACTTGATTTTACTTCC[A/G]GAAAAGCAGTGATTATAGAATTGCT
Phenotype
MIM: 603839
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM126B PubMed Links

Gene Details

Gene
FAM126B
Gene Name
family with sequence similarity 126 member B
There are no transcripts associated with this gene.

Gene
NDUFB3
Gene Name
NADH:ubiquinone oxidoreductase subunit B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257102.1 Intron NP_001244031.1
NM_002491.2 Intron NP_002482.1
XM_011511230.2 Intron XP_011509532.1
XM_017004186.1 Intron XP_016859675.1

View Full Product Details