Product Details

SNP ID
rs55672388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218134844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGAGTGACAGCTTTGAAGATTTC[C/T]GGAAAGGTGAAGATCTTAGTAATTA
Phenotype
MIM: 146928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXCR2 PubMed Links

Gene Details

Gene
CXCR2
Gene Name
C-X-C motif chemokine receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168298.1 124 Missense Mutation CGG,TGG R15W NP_001161770.1
NM_001557.3 124 Missense Mutation CGG,TGG R15W NP_001548.1
XM_005246530.3 124 Intron XP_005246587.1
XM_017003990.1 124 Missense Mutation CGG,TGG R15W XP_016859479.1
XM_017003991.1 124 Missense Mutation CGG,TGG R15W XP_016859480.1
XM_017003992.1 124 Missense Mutation CGG,TGG R15W XP_016859481.1

View Full Product Details