Product Details

SNP ID
rs55716377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222432604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAGTCTGAATGCAAGACCCAGA[A/C]ATAAATGTTAGAGTTCTGACAAACC
Phenotype
MIM: 612827
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SGPP2 PubMed Links

Gene Details

Gene
SGPP2
Gene Name
sphingosine-1-phosphate phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320833.1 Intron NP_001307762.1
NM_001320834.1 Intron NP_001307763.1
NM_152386.3 Intron NP_689599.2
XM_017003355.1 Intron XP_016858844.1

View Full Product Details