Product Details

SNP ID
rs60624480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241357703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCCTTGTTGCAGATTCTCATTAG[A/G]TGGTTCACTGTTAACAAACTAAATG
Phenotype
MIM: 601506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FARP2 PubMed Links

Gene Details

Gene
FARP2
Gene Name
FERM, ARH/RhoGEF and pleckstrin domain protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282983.1 Intron NP_001269912.1
NM_001282984.1 Intron NP_001269913.1
NM_014808.3 Intron NP_055623.1
XM_005247050.3 Intron XP_005247107.1
XM_011512233.1 Intron XP_011510535.1
XM_011512234.1 Intron XP_011510536.1
XM_011512235.1 Intron XP_011510537.1
XM_011512236.1 Intron XP_011510538.1
XM_017005416.1 Intron XP_016860905.1
XM_017005417.1 Intron XP_016860906.1
XM_017005418.1 Intron XP_016860907.1
XM_017005419.1 Intron XP_016860908.1
XM_017005420.1 Intron XP_016860909.1
Gene
SEPT2
Gene Name
septin 2
There are no transcripts associated with this gene.

View Full Product Details