Product Details

SNP ID
rs62187812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240718708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCACCCACCTTGGGTGACTCC[A/G]GAGGAGGGGACATGGCTGTGGCCAT
Phenotype
MIM: 601255
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF1A PubMed Links

Gene Details

Gene
KIF1A
Gene Name
kinesin family member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244008.1 Intron NP_001230937.1
NM_001320705.1 Intron NP_001307634.1
XM_005247022.1 Intron XP_005247079.1
XM_005247024.1 Intron XP_005247081.1
XM_005247027.1 Intron XP_005247084.1
XM_005247028.2 Intron XP_005247085.1
XM_006712605.1 Intron XP_006712668.1
XM_011511364.1 Intron XP_011509666.1
XM_011511365.1 Intron XP_011509667.1
XM_011511366.2 Intron XP_011509668.1
XM_011511367.1 Intron XP_011509669.1
XM_017004383.1 Intron XP_016859872.1
XM_017004384.1 Intron XP_016859873.1
XM_017004385.1 Intron XP_016859874.1
XM_017004386.1 Intron XP_016859875.1
XM_017004387.1 Intron XP_016859876.1
XM_017004388.1 Intron XP_016859877.1
XM_017004389.1 Intron XP_016859878.1
XM_017004390.1 Intron XP_016859879.1
XM_017004391.1 Intron XP_016859880.1

View Full Product Details