Product Details

SNP ID
rs61793786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132320111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGGGTTAAACTGTGTTTAAAAG[G/T]TCTCTTTAATGCAGTAATCCTAAGT
Phenotype
MIM: 171790
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ACPP PubMed Links

Gene Details

Gene
ACPP
Gene Name
acid phosphatase, prostate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099.4 Intron NP_001090.2
NM_001134194.1 Intron NP_001127666.1
NM_001292037.1 Intron NP_001278966.1
XM_011512946.1 Intron XP_011511248.1
XM_011512947.2 Intron XP_011511249.1

View Full Product Details