Product Details
- SNP ID
-
rs56185025
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184571358 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAATGGGTAACATCTGAGTTGGC[A/G]TGGACATCTCATCCAGAAAGTGGGG
- Phenotype
-
MIM: 601839
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPHB3
PubMed Links
Gene Details
- Gene
- EPHB3
- Gene Name
- EPH receptor B3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004443.3 |
596 |
Silent Mutation |
GCA,GCG |
A53A |
NP_004434.2 |
View Full Product Details