Product Details

SNP ID
rs57655409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:42893406 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGCTCTCTGGATTCTGAATGTG[C/T]CAGTATCTGTGGGATAGATGGACTA
Phenotype
MIM: 613283
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRXCR1 PubMed Links

Gene Details

Gene
GRXCR1
Gene Name
glutaredoxin and cysteine rich domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080476.2 141 Missense Mutation GCC,GTC A47V NP_001073945.1

View Full Product Details