Product Details

SNP ID
rs56328578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:16506021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGCAGAATCCAAGGATTAAAC[C/T]CTAGCCCTCGCCAGACACACACATC
Phenotype
MIM: 603450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDB2 PubMed Links

Gene Details

Gene
LDB2
Gene Name
LIM domain binding 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130834.2 Intron NP_001124306.1
NM_001290.4 Intron NP_001281.1
NM_001304434.1 Intron NP_001291363.1
NM_001304435.1 Intron NP_001291364.1
XM_005248197.3 Intron XP_005248254.1
XM_005248201.2 Intron XP_005248258.1
XM_005248202.2 Intron XP_005248259.1
XM_006713975.3 Intron XP_006714038.1
XM_006713976.3 Intron XP_006714039.1
XM_006713977.3 Intron XP_006714040.1
XM_006713978.3 Intron XP_006714041.1
XM_006713979.3 Intron XP_006714042.1
XM_006713980.3 Intron XP_006714043.1
XM_006713981.3 Intron XP_006714044.1
XM_006713982.3 Intron XP_006714045.1
XM_006713983.3 Intron XP_006714046.1
XM_011513905.2 Intron XP_011512207.1
XM_017008812.1 Intron XP_016864301.1
XM_017008813.1 Intron XP_016864302.1
XM_017008814.1 Intron XP_016864303.1
XM_017008815.1 Intron XP_016864304.1
XM_017008816.1 Intron XP_016864305.1
XM_017008817.1 Intron XP_016864306.1
XM_017008818.1 Intron XP_016864307.1
XM_017008819.1 Intron XP_016864308.1
XM_017008820.1 Intron XP_016864309.1
XM_017008821.1 Intron XP_016864310.1
XM_017008822.1 Intron XP_016864311.1

View Full Product Details