Product Details

SNP ID
rs59316994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:45258324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTCTCTGTCCTATAGGAATGA[C/T]GGTAATAACTTGACTATGTACTTTG
Phenotype
MIM: 602780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCN1 PubMed Links

Gene Details

Gene
HCN1
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021072.3 6295 UTR 3 NP_066550.2

View Full Product Details