Product Details

SNP ID
rs56282400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:6599876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCAGACACCAGTGACCAGAAG[A/C]AGCCAGTTTTGCGTGTGAGGGGTGT
Phenotype
MIM: 610916
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NSUN2 PubMed Links

Gene Details

Gene
NSUN2
Gene Name
NOP2/Sun RNA methyltransferase family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193455.1 2735 UTR 3 NP_001180384.1
NM_017755.5 2735 UTR 3 NP_060225.4

View Full Product Details