Product Details

SNP ID
rs56029107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:73499178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGAAAAACTCGCCAAACTGCA[A/G]GCACAAGTGCGCATTGGTGGGAAAG
Phenotype
MIM: 602542
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTF3 PubMed Links

Gene Details

Gene
BTF3
Gene Name
basic transcription factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037637.1 420 Silent Mutation CAA,CAG Q59Q NP_001032726.1
NM_001207.4 420 Silent Mutation CAA,CAG Q15Q NP_001198.2

View Full Product Details