Product Details

SNP ID
rs57806492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:984543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCTGGCCGACCGGCCTCCCAT[C/T]GCCATCTTCCAGCTCATCCAGCGAG
Phenotype
MIM: 610429 MIM: 615967
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C7orf50 PubMed Links

Gene Details

Gene
C7orf50
Gene Name
chromosome 7 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134395.1 319 Intron NP_001127867.1
NM_001134396.1 319 Intron NP_001127868.1
NM_001318252.1 319 Intron NP_001305181.1
NM_032350.5 319 Intron NP_115726.1
XM_006715793.2 319 Intron XP_006715856.1
XM_011515580.2 319 Intron XP_011513882.1
XM_011515581.2 319 Intron XP_011513883.1
XM_011515582.2 319 Intron XP_011513884.1
XM_011515583.2 319 Intron XP_011513885.1
XM_011515584.2 319 Intron XP_011513886.1
XM_017012719.1 319 Intron XP_016868208.1
XM_017012720.1 319 Intron XP_016868209.1
XM_017012721.1 319 Intron XP_016868210.1
XM_017012722.1 319 Intron XP_016868211.1
Gene
COX19
Gene Name
COX19, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.

Gene
CYP2W1
Gene Name
cytochrome P450 family 2 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017781.2 319 Silent Mutation ATC,ATT I102I NP_060251.2
XM_011515440.2 319 Silent Mutation ATC,ATT I102I XP_011513742.1
XM_011515441.2 319 Silent Mutation ATC,ATT I102I XP_011513743.1

View Full Product Details