Product Details

SNP ID
rs61054943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142767785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTGTGCTCAGGGCTCCTTCCT[C/T]ACCCCCACCCAAGCATGAGGGCCTC
Phenotype
MIM: 606110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYNX1 PubMed Links

Gene Details

Gene
LYNX1
Gene Name
Ly6/neurotoxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023946.3 Intron NP_076435.1
NM_177457.4 Intron NP_803252.1
NM_177458.1 Intron NP_803253.1
NM_177476.3 Intron NP_803429.1
NM_177477.3 Intron NP_803430.1

View Full Product Details