Product Details

SNP ID
rs56305287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108868613 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGTAAGATGTTACAAAATAATA[A/G]CTGTCAGCAAAGGAGGGCTTATAAT
Phenotype
MIM: 604303 MIM: 603722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTL7A PubMed Links

Gene Details

Gene
ACTL7A
Gene Name
actin like 7A
There are no transcripts associated with this gene.

Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318360.1 4880 UTR 3 NP_001305289.1
NM_003640.4 4880 UTR 3 NP_003631.2
XM_011519136.1 4880 UTR 3 XP_011517438.1
XM_011519137.1 4880 UTR 3 XP_011517439.1
XM_017015238.1 4880 Intron XP_016870727.1

View Full Product Details