Product Details
- SNP ID
-
rs58894130
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:123381197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGGTGCCGAGGAGAGGCAACCGC[C/G]GGGTGGGATGGGCACTCAGGAACTG
- Phenotype
-
MIM: 609720
MIM: 613633
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CRB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77969061] are located under a probe and SNP(s) [rs139826267] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CRB2
- Gene Name
- crumbs 2, cell polarity complex component
- Gene
- DENND1A
- Gene Name
- DENN domain containing 1A
View Full Product Details