Product Details

SNP ID
rs55758160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128922736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAATCACAAAAGGCTACCTCGT[A/G]GGAATTAGTGTGCAACAGCCTTTTA
Phenotype
MIM: 608360
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC8A PubMed Links

Gene Details

Gene
LRRC8A
Gene Name
leucine rich repeat containing 8 family member A
There are no transcripts associated with this gene.

Gene
PHYHD1
Gene Name
phytanoyl-CoA dioxygenase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100876.1 Intron NP_001094346.1
NM_001100877.1 Intron NP_001094347.1
NM_174933.3 Intron NP_777593.2

View Full Product Details