Product Details
- SNP ID
-
rs60385020
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:129042001 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGTACCCCAGGCTAGGTCCTAGT[G/T]CAGTGCCGGGCACAGCTGGTGCTTG
- Phenotype
-
MIM: 616774
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MIGA2
PubMed Links
Gene Details
- Gene
- MIGA2
- Gene Name
- mitoguardin 2
View Full Product Details