Product Details

SNP ID
rs61755667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:67925146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATACTTCCCAAAAAATTTTTAT[C/T]GTAGGATGTTCTGCCCAATATTCCC
Phenotype
MIM: 609248 MIM: 604479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HERC4 PubMed Links

Gene Details

Gene
HERC4
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278185.1 3265 Silent Mutation NP_001265114.1
NM_001278186.1 3265 Silent Mutation NP_001265115.1
NM_001278187.1 3265 Intron NP_001265116.1
NM_015601.3 3265 Silent Mutation NP_056416.2
NM_022079.2 3265 Silent Mutation NP_071362.1
XM_011539592.2 3265 Silent Mutation XP_011537894.1
XM_011539593.2 3265 Silent Mutation XP_011537895.1
XM_011539594.2 3265 Silent Mutation XP_011537896.1
XM_011539595.2 3265 Silent Mutation XP_011537897.1
XM_011539596.2 3265 Silent Mutation XP_011537898.1
XM_011539597.2 3265 Silent Mutation XP_011537899.1
XM_017016041.1 3265 Silent Mutation XP_016871530.1
Gene
SIRT1
Gene Name
sirtuin 1
There are no transcripts associated with this gene.

View Full Product Details