Product Details

SNP ID
rs11557024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79349094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACAGCTGAAGGCAGATGTCGTC[A/C]CAAAGACAGCTGGTAAGACAGGGCC
Phenotype
MIM: 604486
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPIF PubMed Links

Gene Details

Gene
PPIF
Gene Name
peptidylprolyl isomerase F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005729.3 297 Missense Mutation NP_005720.1
XM_005269379.2 297 Missense Mutation XP_005269436.1

View Full Product Details