Product Details

SNP ID
rs61880539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:20675073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGGCTGCCCGCTCCACATGTCT[A/G]GGTCTCCTTGGACCAGTGGACCAGC
Phenotype
MIM: 602319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NELL1 PubMed Links

Gene Details

Gene
NELL1
Gene Name
neural EGFL like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288713.1 Intron NP_001275642.1
NM_001288714.1 Intron NP_001275643.1
NM_006157.4 Intron NP_006148.2
NM_201551.2 Intron NP_963845.1

View Full Product Details