Product Details

SNP ID
rs61927807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:21356760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAATAATAGTAGCTTAATTCT[C/T]ATGCAGTCCAAGTTCTAGTAAGGTA
Phenotype
MIM: 147940 MIM: 602883
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IAPP PubMed Links

Gene Details

Gene
IAPP
Gene Name
islet amyloid polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000415.2 Intron NP_000406.1
XM_017019253.1 Intron XP_016874742.1
Gene
SLCO1A2
Gene Name
solute carrier organic anion transporter family member 1A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021094.3 Intron NP_066580.1
NM_134431.3 Intron NP_602307.1
XM_005253474.3 Intron XP_005253531.1
XM_005253477.3 Intron XP_005253534.1
XM_011520818.1 Intron XP_011519120.1
XM_011520819.1 Intron XP_011519121.1
XM_011520820.2 Intron XP_011519122.1
XM_011520821.2 Intron XP_011519123.1
XM_017019849.1 Intron XP_016875338.1
XM_017019850.1 Intron XP_016875339.1

View Full Product Details