Product Details

SNP ID
rs61913639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9596900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATAAAATATTGAAAATAACCC[A/C]CAGAAAGCAATATAAAAATCAAGAG
Phenotype
MIM: 602890
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLRB1 PubMed Links

Gene Details

Gene
KLRB1
Gene Name
killer cell lectin like receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002258.2 Intron NP_002249.1

View Full Product Details