Product Details

SNP ID
rs61916315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6831328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACCAGGCCCAGCTGGGAGAGCC[A/G]AGACCTGGCCTCGGACCCAGAGAGG
Phenotype
MIM: 139130 MIM: 610342
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNB3 PubMed Links

Gene Details

Gene
GNB3
Gene Name
G protein subunit beta 3
There are no transcripts associated with this gene.

Gene
GPR162
Gene Name
G protein-coupled receptor 162
There are no transcripts associated with this gene.

Gene
P3H3
Gene Name
prolyl 3-hydroxylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014262.4 1166 Silent Mutation CCA,CCG P366P NP_055077.2

View Full Product Details