Product Details
- SNP ID
-
hCV89193420
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:40405970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGGGCTAAGCGAGGAGCAGAGGC[A/G]GGTGAGGAGACTGACCCCCTTCCTG
- Phenotype
-
MIM: 607036
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IVD
PubMed Links
Gene Details
- Gene
- IVD
- Gene Name
- isovaleryl-CoA dehydrogenase
There are no transcripts associated with this gene.
View Full Product Details