Product Details

SNP ID
rs60181666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52757803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACCAACCTGGGCTTTTTGGGTG[C/T]GTTGCCTCTATCCTTCCCATGTTCT
Phenotype
MIM: 604164
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ONECUT1 PubMed Links

Gene Details

Gene
ONECUT1
Gene Name
one cut homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004498.2 1278 Missense Mutation ACA,GCA T384A NP_004489.1
XM_011521487.2 1278 Intron XP_011519789.1

View Full Product Details