Product Details
- SNP ID
-
rs59705963
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:68284542 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAGTGGAGGTATATCATATCTAG[G/T]AAAGTACATGAATCTTGTGTACAAT
- Phenotype
-
MIM: 613539
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FEM1B
PubMed Links
Gene Details
- Gene
- FEM1B
- Gene Name
- fem-1 homolog B
View Full Product Details