Product Details

SNP ID
rs60958974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78627412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAAGAACTACACCAGAAGAATA[C/T]CTCATGTCCTATATTCCATTTGTTT
Phenotype
MIM: 118503 MIM: 118509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNA3 PubMed Links

Gene Details

Gene
CHRNA3
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
There are no transcripts associated with this gene.

Gene
CHRNB4
Gene Name
cholinergic receptor nicotinic beta 4 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000750.3 Intron NP_000741.1
NM_001256567.1 Intron NP_001243496.1
XM_011521186.2 Intron XP_011519488.1
XM_011521187.2 Intron XP_011519489.1
XM_011521190.2 Intron XP_011519492.1
XM_011521191.2 Intron XP_011519493.1
XM_011521192.2 Intron XP_011519494.1
XM_017021885.1 Intron XP_016877374.1
XM_017021886.1 Intron XP_016877375.1
XM_017021887.1 Intron XP_016877376.1
XM_017021888.1 Intron XP_016877377.1
XM_017021889.1 Intron XP_016877378.1

View Full Product Details