Product Details

SNP ID
rs56273424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90187549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCACTGCAGCATAATGAGGTC[C/T]TAATAAGAATCTGAGCAGAGTGCAG
Phenotype
MIM: 617029
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA4B PubMed Links

Gene Details

Gene
SEMA4B
Gene Name
semaphorin 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324029.1 Intron NP_001310958.1
NM_001324030.1 Intron NP_001310959.1
NM_001324031.1 Intron NP_001310960.1
NM_001324032.1 Intron NP_001310961.1
NM_001324033.1 Intron NP_001310962.1
NM_001324034.1 Intron NP_001310963.1
NM_020210.3 Intron NP_064595.2
NM_198925.2 Intron NP_945119.1

View Full Product Details