Product Details

SNP ID
rs55649477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3724837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCCACAGGCCAGGACTCTTGTC[A/T]AGAGAGAGGGCTTCAAGAGGCCGTC
Phenotype
MIM: 609240 MIM: 604682
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GSG2 PubMed Links

Gene Details

Gene
GSG2
Gene Name
germ cell associated 2, haspin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031965.2 935 Missense Mutation CAA,CTA Q301L NP_114171.2
Gene
ITGAE
Gene Name
integrin subunit alpha E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002208.4 935 Intron NP_002199.3
XM_011523823.2 935 Intron XP_011522125.1
XM_011523825.2 935 Intron XP_011522127.1
XM_011523827.2 935 Intron XP_011522129.1
XM_011523828.2 935 Intron XP_011522130.1
XM_017024586.1 935 Intron XP_016880075.1
XM_017024587.1 935 Intron XP_016880076.1

View Full Product Details