Product Details

SNP ID
rs58212531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8100234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGAAATCTGAAAGCTTGTTTG[C/T]AATCTTGGAAGCCAGCCACCGTGCT
Phenotype
MIM: 607206
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOXE3 PubMed Links

Gene Details

Gene
ALOXE3
Gene Name
arachidonate lipoxygenase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165960.1 Intron NP_001159432.1
NM_021628.2 Intron NP_067641.2
XM_017024921.1 Intron XP_016880410.1
XM_017024922.1 Intron XP_016880411.1
XM_017024923.1 Intron XP_016880412.1
XM_017024924.1 Intron XP_016880413.1
XM_017024925.1 Intron XP_016880414.1

View Full Product Details