Product Details

SNP ID
rs55913274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28042936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGGCTTACAATGGCGGTACATC[C/T]GCAGCAGCAGCAGGTCACCACCACC
Phenotype
MIM: 609476
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NLK PubMed Links

Gene Details

Gene
NLK
Gene Name
nemo like kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016231.4 290 Silent Mutation TCC,TCT S21S NP_057315.3
XM_005257988.2 290 Silent Mutation TCC,TCT S21S XP_005258045.1

View Full Product Details