Product Details
- SNP ID
-
hCV89423646
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:39972539 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTGTGGATCAGAATACAGCCAT[C/T]GTGGGCTCTACCACAGGTGTGCTAA
- Phenotype
-
MIM: 602707
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSMC4
PubMed Links
Gene Details
- Gene
- PSMC4
- Gene Name
- proteasome 26S subunit, ATPase 4
There are no transcripts associated with this gene.
View Full Product Details