Product Details

SNP ID
rs61435346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40481162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTTCTGGTGGTTTACACCCAC[A/G]CAGGAGTGAAGCTGTCCAATCATAG
Phenotype
MIM: 606214
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPTBN4 PubMed Links

Gene Details

Gene
SPTBN4
Gene Name
spectrin beta, non-erythrocytic 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020971.2 Intron NP_066022.2
NM_025213.2 Intron NP_079489.2
XM_011527173.2 Intron XP_011525475.1
XM_017027049.1 Intron XP_016882538.1
XM_017027050.1 Intron XP_016882539.1
XM_017027051.1 Intron XP_016882540.1
XM_017027052.1 Intron XP_016882541.1

View Full Product Details