Product Details

SNP ID
rs61764024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48326834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTGCTCCCCCTCCCCAGGCTT[C/G]CACTGCAGCCATGTCACTCCTCTTG
Phenotype
MIM: 615038 MIM: 602335
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC114 PubMed Links

Gene Details

Gene
CCDC114
Gene Name
coiled-coil domain containing 114
There are no transcripts associated with this gene.

Gene
EMP3
Gene Name
epithelial membrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313905.1 175 UTR 5 NP_001300834.1
NM_001425.2 175 UTR 5 NP_001416.1
XM_011526605.2 175 UTR 5 XP_011524907.1
Gene
TMEM143
Gene Name
transmembrane protein 143
There are no transcripts associated with this gene.

View Full Product Details