Product Details

SNP ID
rs62129562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55658072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCAGACATTCACATTGCATC[A/G]CAAAAACGTGAGACAAAACACTGTG
Phenotype
MIM: 613478 MIM: 191318
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC106 PubMed Links

Gene Details

Gene
CCDC106
Gene Name
coiled-coil domain containing 106
There are no transcripts associated with this gene.

Gene
U2AF2
Gene Name
U2 small nuclear RNA auxiliary factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012478.1 Intron NP_001012496.1
NM_007279.2 Intron NP_009210.1
XM_006722994.2 Intron XP_006723057.1
XM_011526410.1 Intron XP_011524712.1

View Full Product Details