Product Details

SNP ID
hCV89480805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62807541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCATCTCTTTTCTTCCCAGTCCA[G/T]AATGACAGGGTCCAGAAACTGGCGA
Phenotype
MIM: 120270 MIM: 611157 MIM: 606459
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COL9A3 PubMed Links

Gene Details

Gene
COL9A3
Gene Name
collagen type IX alpha 3
There are no transcripts associated with this gene.

Gene
MRGBP
Gene Name
MRG/MORF4L binding protein
There are no transcripts associated with this gene.

Gene
OGFR
Gene Name
opioid growth factor receptor
There are no transcripts associated with this gene.

Gene
OGFR-AS1
Gene Name
OGFR antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details