Product Details

SNP ID
rs62192710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:1896950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCGAGTTCCTGCCCTGCCATCC[G/T]TCCTGGCCTGGAAATGCACAAGTGT
Phenotype
MIM: 602461
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SIRPA PubMed Links

Gene Details

Gene
SIRPA
Gene Name
signal regulatory protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040022.1 Intron NP_001035111.1
NM_001040023.1 Intron NP_001035112.1
NM_080792.2 Intron NP_542970.1
XM_005260669.3 Intron XP_005260726.1
XM_005260670.3 Intron XP_005260727.1
XM_006723545.3 Intron XP_006723608.1
XM_011529173.2 Intron XP_011527475.1

View Full Product Details