Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
XM_017028182.1 | Intron | XP_016883671.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001323679.1 | Intron | NP_001310608.1 | |||
NM_001323680.1 | Intron | NP_001310609.1 | |||
NM_001323681.1 | Intron | NP_001310610.1 | |||
NM_001323682.1 | Intron | NP_001310611.1 | |||
NM_001323683.1 | Intron | NP_001310612.1 | |||
NM_001323684.1 | Intron | NP_001310613.1 | |||
NM_001323685.1 | Intron | NP_001310614.1 | |||
NM_024958.2 | Intron | NP_079234.1 | |||
XM_006723630.3 | Intron | XP_006723693.1 | |||
XM_011529360.1 | Intron | XP_011527662.1 | |||
XM_011529362.2 | Intron | XP_011527664.1 | |||
XM_011529363.2 | Intron | XP_011527665.1 | |||
XM_017028073.1 | Intron | XP_016883562.1 | |||
XM_017028074.1 | Intron | XP_016883563.1 | |||
XM_017028075.1 | Intron | XP_016883564.1 | |||
XM_017028076.1 | Intron | XP_016883565.1 |